Assay Details
Target Gene Details
Entrez Gene ID: | 101928977 |
Gene Name: | uncharacterized LOC101928977 |
Gene Aliases: |
- |
Location: |
Chr.1:115922573-115977276 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928977 | NR_125970.1 | |||
| DA284216.1 | ||||
| DA297063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760079 | Chr.1:115942312 - 115948700 on Build GRCh38 | Loss |
|
| esv27976 | Chr.1:115592696 - 116135006 on Build GRCh38 | Loss |
|
| esv3587294 | Chr.1:115861399 - 115956767 on Build GRCh38 | Loss |
|

Genomic Map