Assay Details
Target Gene Details
Entrez Gene ID: | 8880 |
Gene Name: | far upstream element binding protein 1 |
Gene Aliases: |
FBP, FUBP, hDH V |
Location: |
Chr.1:77944055-77979494 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FUBP1 | XM_011542391.2 | 22 | 2206 | XP_011540693.1 |
| XM_011542392.2 | 22 | 2237 | XP_011540694.1 | |
| XM_011542393.2 | 21 | 2177 | XP_011540695.1 | |
| XM_017002739.1 | 23 | 2228 | XP_016858228.1 | |
| XM_017002740.1 | 21 | 4069 | XP_016858229.1 | |
| XM_017002741.1 | 22 | 2221 | XP_016858230.1 | |
| XM_017002742.1 | 21 | 5322 | XP_016858231.1 | |
| XM_017002743.1 | 20 | 4041 | XP_016858232.1 | |
| XM_017002744.1 | 20 | 5263 | XP_016858233.1 | |
| AB209366.1 | 13 | 5086 | BAD92603.1 | |
| AL049951.1 | 1 | 218 | ||
| BX538272.1 | 3 | 1796 |
Target Gene Details
Entrez Gene ID: | 91624 |
Gene Name: | nexilin F-actin binding protein |
Gene Aliases: |
CMH20, NELIN |
Location: |
Chr.1:77888515-77946329 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEXN | XM_005271322.3 | 14 | 4851 | XP_005271379.1 |
| XM_005271325.3 | 13 | 4629 | XP_005271382.1 |
More Information
Set Membership: |
|

Genomic Map