Assay Details
Target Gene Details
Entrez Gene ID: | 9447 |
Gene Name: | absent in melanoma 2 |
Gene Aliases: |
PYHIN4 |
Location: |
Chr.1:159059226-159132351 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIM2 | XM_017002848.1 | XP_016858337.1 |
More Information
Set Membership: |
|

Genomic Map