Assay Details
Target Gene Details
Entrez Gene ID: | 4921 |
Gene Name: | discoidin domain receptor tyrosine kinase 2 |
Gene Aliases: |
MIG20a, NTRKR3, TKT, TYRO10 |
Location: |
Chr.1:162631213-162783524 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDR2 | NM_001014796.1 | NP_001014796.1 | ||
| NM_006182.2 | NP_006173.2 | |||
| XM_006711344.3 | XP_006711407.1 | |||
| XM_011509586.2 | XP_011507888.1 | |||
| XM_011509587.2 | XP_011507889.1 | |||
| XM_011509588.2 | XP_011507890.1 | |||
| AK095975.1 | ||||
| AK225063.1 | ||||
| AK225196.1 | ||||
| AK314388.1 | ||||
| BC052998.2 | AAH52998.1 | |||
| BX640899.1 | CAE45946.1 | |||
| CF994640.1 | ||||
| X74764.1 | CAA52777.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516482 | Chr.1:162138329 - 162905405 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map