Assay Details
Target Gene Details
Entrez Gene ID: | 115350 |
Gene Name: | Fc receptor like 1 |
Gene Aliases: |
CD307a, FCRH1, IFGP1, IRTA5 |
Location: |
Chr.1:157792249-157820150 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRL1 | XM_005244866.2 | 11 | 3557 | XP_005244923.1 |
| XM_005244867.2 | 10 | 3450 | XP_005244924.1 | |
| XM_005244869.3 | 9 | 3292 | XP_005244926.1 | |
| XM_011509125.1 | 12 | 3565 | XP_011507427.1 | |
| XM_011509126.1 | 12 | 3562 | XP_011507428.1 | |
| XM_011509127.2 | 10 | 3559 | XP_011507429.1 | |
| XM_011509128.1 | 11 | 3569 | XP_011507430.1 | |
| XM_011509129.1 | 11 | 3566 | XP_011507431.1 | |
| XM_011509130.1 | 11 | 3554 | XP_011507432.1 | |
| XM_011509133.2 | 11 | 3421 | XP_011507435.1 | |
| XM_011509134.2 | 10 | 3396 | XP_011507436.1 | |
| XM_011509135.2 | 10 | 3313 | XP_011507437.1 | |
| XM_011509137.2 | 8 | 2941 | XP_011507439.1 | |
| XM_017000227.1 | 10 | 5563 | XP_016855716.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587657 | Chr.1:157793395 - 157801763 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map