Assay Details
Target Gene Details
Entrez Gene ID: | 55356 |
Gene Name: | solute carrier family 22 member 15 |
Gene Aliases: |
FLIPT1, PRO34686 |
Location: |
Chr.1:115976498-116070054 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A15 | NM_018420.2 | NP_060890.2 | ||
| XM_005271004.3 | XP_005271061.1 | |||
| AY145501.1 | AAN52927.1 | |||
| AY358258.1 | AAQ88625.1 | |||
| BC026358.1 | AAH26358.1 | |||
| KU178698.1 | ||||
| KU178699.1 | ||||
| KU178700.1 | ||||
| KU178701.1 | ||||
| KU178702.1 | ||||
| KU178703.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27976 | Chr.1:115592696 - 116135006 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map