Assay Details
Target Gene Details
Entrez Gene ID: | 10076 |
Gene Name: | protein tyrosine phosphatase, receptor type U |
Gene Aliases: |
FMI, PCP-2, PTP, PTP-J, PTP-PI, PTP-RO, PTPPSI, PTPRO, PTPU2, R-PTP-PSI, R-PTP-U, hPTP-J |
Location: |
Chr.1:29236516-29326813 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRU | NM_001195001.1 | NP_001181930.1 | ||
| NM_005704.4 | NP_005695.3 | |||
| NM_133177.3 | NP_573438.3 | |||
| NM_133178.3 | NP_573439.2 | |||
| XM_006710269.2 | XP_006710332.1 | |||
| XM_016999992.1 | XP_016855481.1 | |||
| XM_016999993.1 | XP_016855482.1 | |||
| XM_016999994.1 | XP_016855483.1 | |||
| BC146655.1 | ||||
| U71075.1 | AAC51938.1 | |||
| U73727.1 | AAB51343.1 | |||
| X95712.1 | CAA65016.1 | |||
| X97198.1 | CAA65832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834546 | Chr.1:29245770 - 29433789 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77830010] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map