Assay Details
Target Gene Details
Entrez Gene ID: | 3932 |
Gene Name: | LCK proto-oncogene, Src family tyrosine kinase |
Gene Aliases: |
IMD22, LSK, YT16, p56lck, pp58lck |
Location: |
Chr.1:32251239-32286167 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCK | NM_001042771.2 | 7 | 686 | NP_001036236.1 |
| NM_005356.4 | 7 | 703 | NP_005347.3 | |
| XM_011541453.2 | 6 | 877 | XP_011539755.1 | |
| XM_011541454.2 | 6 | 858 | XP_011539756.1 | |
| XM_011541455.2 | 7 | 703 | XP_011539757.1 | |
| AF228313.1 | 6 | 526 | AAF34794.1 | |
| AJ865079.1 | 5 | 739 | CAI23831.1 | |
| AK098027.1 | 7 | 500 | ||
| AK303073.1 | 6 | 756 | ||
| BC013200.1 | 7 | 665 | AAH13200.1 | |
| DB129997.1 | ||||
| KU178161.1 | 6 | 565 | ||
| KU178162.1 | 6 | 565 | ||
| KU178163.1 | 6 | 565 | ||
| M36881.1 | 7 | 616 | AAA59502.1 | |
| U07236.1 | 7 | 591 | AAA18225.1 | |
| X05027.1 | 7 | 657 | CAA28691.1 | |
| X13529.1 | 7 | 616 | CAA31884.1 |
More Information
Set Membership: |
|

Genomic Map