Assay Details
Target Gene Details
Entrez Gene ID: | 101928650 |
Gene Name: | uncharacterized LOC101928650 |
Gene Aliases: |
- |
Location: |
Chr.1:170461317-170532609 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928650 | NR_125958.1 | |||
| AK096329.1 | ||||
| HY028703.1 | ||||
| HY035829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587898 | Chr.1:170490253 - 170492434 on Build GRCh38 | Loss |
|

Genomic Map