Assay Details
Target Gene Details
Entrez Gene ID: | 9447 |
Gene Name: | absent in melanoma 2 |
Gene Aliases: |
PYHIN4 |
Location: |
Chr.1:159059226-159132351 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIM2 | NM_004833.1 | NP_004824.1 | ||
| XM_005245616.4 | XP_005245673.1 | |||
| XM_017002848.1 | XP_016858337.1 | |||
| AF024714.1 | AAB81613.1 | |||
| AK292042.1 | ||||
| BC010940.1 | AAH10940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv946445 | Chr.1:159065253 - 159066111 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map