Assay Details
Target Gene Details
Entrez Gene ID: | 5792 |
Gene Name: | protein tyrosine phosphatase, receptor type F |
Gene Aliases: |
BNAH2, LAR |
Location: |
Chr.1:43522238-43623672 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv157n100 | Chr.1:43525952 - 43606749 on Build GRCh38 | Gain |
|
| dgv158n100 | Chr.1:43559459 - 43620498 on Build GRCh38 | Gain |
|
| dgv82e59 | Chr.1:43558994 - 43561092 on Build GRCh38 | Duplication |
|
| nsv519570 | Chr.1:43539609 - 43606395 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs68191270] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map