Assay Details
Target Gene Details
Entrez Gene ID: | 3284 |
Gene Name: | hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2 |
Gene Aliases: |
HSD3B, HSDB, SDR11E2 |
Location: |
Chr.1:119414931-119423039 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD3B2 | NM_000198.3 | NP_000189.1 | ||
| NM_001166120.1 | NP_001159592.1 | |||
| AK222997.1 | BAD96717.1 | |||
| AK313413.1 | ||||
| AV705363.1 | ||||
| BC038419.1 | AAH38419.1 | |||
| BC131488.1 | ||||
| BP197992.1 | ||||
| CF596786.1 | ||||
| M67466.1 | AAA36016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763647 | Chr.1:119386819 - 119612175 on Build GRCh38 | Gain+Loss |
|
| nsv2599 | Chr.1:119392400 - 119425125 on Build GRCh38 | Deletion |
|
| esv34089 | Chr.1:119209489 - 119530338 on Build GRCh38 | Loss |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
| nsv1004662 | Chr.1:119304824 - 119612175 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map