Assay Details
Target Gene Details
Entrez Gene ID: | 55160 |
Gene Name: | Rho guanine nucleotide exchange factor 10 like |
Gene Aliases: |
GrinchGEF |
Location: |
Chr.1:17520197-17697875 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF10L | NM_018125.3 | NP_060595.3 | ||
| NR_137287.1 | ||||
| NR_137288.1 | ||||
| XM_005245923.2 | XP_005245980.2 | |||
| XM_005245925.2 | XP_005245982.2 | |||
| XM_006710728.1 | XP_006710791.1 | |||
| XM_006710729.1 | XP_006710792.1 | |||
| XM_006710731.1 | XP_006710794.1 | |||
| XM_017001617.1 | XP_016857106.1 | |||
| AB046846.1 | ||||
| AK129959.1 | ||||
| AK311076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585371 | Chr.1:17541780 - 17543451 on Build GRCh38 | Loss |
|
| nsv834302 | Chr.1:17531435 - 17713452 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map