Assay Details
Target Gene Details
Entrez Gene ID: | 515 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1 |
Gene Aliases: |
PIG47 |
Location: |
Chr.1:111449121-111461918 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5F1 | NM_001688.4 | NP_001679.2 | ||
| AK223018.1 | BAD96738.1 | |||
| AK291077.1 | ||||
| AK308266.1 | ||||
| AK311287.1 | ||||
| AK313541.1 | ||||
| AY871272.1 | ||||
| BC005366.1 | AAH05366.1 | |||
| BC005960.1 | AAH05960.1 | |||
| BC016350.1 | AAH16350.1 | |||
| BF237506.1 | ||||
| X60221.1 | CAA42782.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547535 | Chr.1:111195294 - 111519590 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76903514] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map