Assay Details
Target Gene Details
Entrez Gene ID: | 55509 |
Gene Name: | basic leucine zipper ATF-like transcription factor 3 |
Gene Aliases: |
JDP1, JUNDM1, SNFT |
Location: |
Chr.1:212686417-212699985 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BATF3 | NM_018664.2 | NP_061134.1 | ||
| XM_017001683.1 | 3 | 546 | XP_016857172.1 | |
| AF255346.1 | AAF73966.1 | |||
| AK093671.1 | ||||
| BC117489.1 | ||||
| BC117491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549139 | Chr.1:212158952 - 212703560 on Build GRCh38 | Gain |
|
| nsv4465 | Chr.1:212665368 - 212699073 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map