Assay Details
Target Gene Details
Entrez Gene ID: | 64744 |
Gene Name: | small ArfGAP2 |
Gene Aliases: |
SMAP1L |
Location: |
Chr.1:40373706-40423326 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAP2 | NM_001198978.1 | NP_001185907.1 | ||
| NM_001198979.1 | NP_001185908.1 | |||
| NM_022733.2 | NP_073570.1 | |||
| XM_011541960.1 | XP_011540262.1 | |||
| XM_017002093.1 | XP_016857582.1 | |||
| AK303975.1 | ||||
| AK308280.1 | ||||
| AK313105.1 | ||||
| AL133206.1 | CAB61580.1 | |||
| AL137764.1 | CAB70912.1 | |||
| BC021133.1 | AAH21133.1 | |||
| DC411665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950697 | Chr.1:40392029 - 40404928 on Build GRCh38 | Duplication |
|
| esv3585803 | Chr.1:40375034 - 40406101 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map