Assay Details
Target Gene Details
Entrez Gene ID: | 23499 |
Gene Name: | microtubule-actin crosslinking factor 1 |
Gene Aliases: |
ABP620, ACF7, MACF, OFC4 |
Location: |
Chr.1:39084167-39487138 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MACF1 | NM_012090.5 | NP_036222.3 | ||
| XM_006710526.3 | XP_006710589.1 | |||
| XM_006710535.3 | XP_006710598.1 | |||
| XM_017000875.1 | XP_016856364.1 | |||
| XM_017000876.1 | XP_016856365.1 | |||
| XM_017000881.1 | XP_016856370.1 | |||
| AB029290.1 | BAA83821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585784 | Chr.1:39110284 - 39112127 on Build GRCh38 | Loss |
|
| nsv515793 | Chr.1:39105539 - 39112860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map