Assay Details
Target Gene Details
Entrez Gene ID: | 9926 |
Gene Name: | lysophosphatidylglycerol acyltransferase 1 |
Gene Aliases: |
FAM34A, FAM34A1, NET8 |
Location: |
Chr.1:211743457-211830772 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPGAT1 | NM_001320808.1 | 8 | 7739 | NP_001307737.1 |
| NM_014873.2 | 8 | 7150 | NP_055688.1 | |
| XM_011510228.2 | 9 | 7187 | XP_011508530.1 | |
| XM_011510229.2 | 8 | 7070 | XP_011508531.1 | |
| XM_017003004.1 | 7 | 8302 | XP_016858493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3588735 | Chr.1:211709170 - 211744936 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78359634] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map