Assay Details
Target Gene Details
Entrez Gene ID: | 6402 |
Gene Name: | selectin L |
Gene Aliases: |
CD62L, LAM1, LECAM1, LEU8, LNHR, LSEL, LYAM1, PLNHR, TQ1 |
Location: |
Chr.1:169690665-169711702 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SELL | NM_000655.4 | NP_000646.2 | ||
| NR_029467.1 | ||||
| AB583235.1 | ||||
| AJ246000.1 | CAB55488.1 | |||
| AK225713.1 | ||||
| AK298705.1 | ||||
| AK312673.1 | ||||
| M25280.1 | AAC63053.1 | |||
| X16070.1 | CAA34203.1 | |||
| X16150.1 | CAA34275.1 | |||
| X17519.1 | CAB43536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009313 | Chr.1:169692158 - 169711032 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map