Assay Details
Target Gene Details
Entrez Gene ID: | 10352 |
Gene Name: | tryptophanyl tRNA synthetase 2, mitochondrial |
Gene Aliases: |
TrpRS |
Location: |
Chr.1:119031216-119140701 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WARS2 | NM_015836.3 | NP_056651.1 | ||
| NM_201263.2 | NP_957715.1 | |||
| XM_005270350.3 | XP_005270407.1 | |||
| XM_011540494.2 | XP_011538796.1 | |||
| XM_011540495.2 | XP_011538797.1 | |||
| XM_017000038.1 | XP_016855527.1 | |||
| XM_017000039.1 | XP_016855528.1 | |||
| XM_017000040.1 | XP_016855529.1 | |||
| XM_017000041.1 | XP_016855530.1 | |||
| XM_017000042.1 | XP_016855531.1 | |||
| AJ242739.1 | CAB63107.1 | |||
| AK223197.1 | BAD96917.1 | |||
| AK296088.1 | ||||
| AK296799.1 | ||||
| AK299547.1 | ||||
| AK300295.1 | ||||
| AK313740.1 | ||||
| BC044575.1 | AAH44575.1 | |||
| BM834499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997707 | Chr.1:118656945 - 119053326 on Build GRCh38 | Gain |
|
| dgv16n21 | Chr.1:119039277 - 119046720 on Build GRCh38 | Loss |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs67639146] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map