Assay Details
Target Gene Details
Entrez Gene ID: | 55105 |
Gene Name: | G-patch domain containing 2 |
Gene Aliases: |
CT110, GPATC2, PPP1R30 |
Location: |
Chr.1:217426992-217631312 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPATCH2 | NM_018040.3 | 10 | 2787 | NP_060510.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832559 | Chr.1:217305459 - 217482303 on Build GRCh38 | Loss |
|
| nsv826631 | Chr.1:217429992 - 217430504 on Build GRCh38 | Gain |
|
| esv2762164 | Chr.1:217216609 - 217569408 on Build GRCh38 | Gain |
|
| nsv832570 | Chr.1:217402639 - 217576576 on Build GRCh38 | Gain |
|
| nsv1008328 | Chr.1:217300400 - 217602339 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map