Assay Details
Target Gene Details
Entrez Gene ID: | 4678 |
Gene Name: | nuclear autoantigenic sperm protein |
Gene Aliases: |
FLB7527, HMDRA1, PRO1999 |
Location: |
Chr.1:45583988-45618906 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NASP | NM_001195193.1 | NP_001182122.1 | ||
| NM_002482.3 | NP_002473.2 | |||
| NM_152298.3 | NP_689511.2 | |||
| XM_005270888.2 | XP_005270945.1 | |||
| XM_005270889.2 | XP_005270946.1 | |||
| XM_011541509.1 | XP_011539811.1 | |||
| XM_017001356.1 | XP_016856845.1 | |||
| AF035191.1 | AAD02006.1 | |||
| AK056161.1 | ||||
| AK092829.1 | 1 | 571 | ||
| AK131249.1 | ||||
| AK222778.1 | BAD96498.1 | |||
| AK222816.1 | BAD96536.1 | |||
| AK291637.1 | ||||
| AK298893.1 | ||||
| AK299584.1 | ||||
| AK308001.1 | ||||
| BC003113.1 | AAH03113.1 | |||
| BC010105.2 | AAH10105.1 | |||
| BC011580.1 | AAH11580.1 | |||
| BC053608.1 | AAH53608.1 | |||
| BT006757.1 | AAP35403.1 | |||
| M97856.1 | AAA36027.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain |
|
| nsv829726 | Chr.1:45431909 - 45637514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map