Assay Details
Target Gene Details
Entrez Gene ID: | 1131 |
Gene Name: | cholinergic receptor muscarinic 3 |
Gene Aliases: |
EGBRS, HM3, PBS |
Location: |
Chr.1:239386565-239915450 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRM3 | XM_005273032.3 | XP_005273089.1 | ||
| XM_011544041.2 | XP_011542343.1 | |||
| XM_011544043.2 | XP_011542345.1 | |||
| XM_011544044.2 | XP_011542346.1 | |||
| XM_011544045.2 | XP_011542347.1 | |||
| XM_011544046.2 | XP_011542348.1 | |||
| XM_011544047.2 | XP_011542349.1 | |||
| XM_017000153.1 | XP_016855642.1 | |||
| XM_017000155.1 | XP_016855644.1 | |||
| XM_017000156.1 | XP_016855645.1 | |||
| XM_017000158.1 | XP_016855647.1 | |||
| XM_017000159.1 | XP_016855648.1 | |||
| XM_017000161.1 | XP_016855650.1 | |||
| XM_017000162.1 | XP_016855651.1 | |||
| AK056349.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575817 | Chr.1:239187428 - 239392293 on Build GRCh38 | Gain |
|
| nsv1002301 | Chr.1:239018955 - 239611093 on Build GRCh38 | Gain |
|
| dgv594n100 | Chr.1:239192375 - 239401382 on Build GRCh38 | Gain |
|
| nsv549435 | Chr.1:239211710 - 239810706 on Build GRCh38 | Gain |
|
| nsv833037 | Chr.1:239358868 - 239536112 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map