Assay Details
Target Gene Details
Entrez Gene ID: | 64388 |
Gene Name: | gremlin 2, DAN family BMP antagonist |
Gene Aliases: |
CKTSF1B2, DAND3, PRDC |
Location: |
Chr.1:240489573-240612372 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GREM2 | NM_022469.3 | NP_071914.3 | ||
| XM_005273226.3 | XP_005273283.1 | |||
| XM_011544249.2 | XP_011542551.1 | |||
| AK024848.1 | BAB15026.1 | |||
| BC046632.1 | AAH46632.1 |
More Information
Set Membership: |
|

Genomic Map