Assay Details
Target Gene Details
Entrez Gene ID: | 6121 |
Gene Name: | RPE65, retinoid isomerohydrolase |
Gene Aliases: |
BCO3, LCA2, RP20, mRPE65, rd12, sRPE65 |
Location: |
Chr.1:68428822-68450322 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPE65 | NM_000329.2 | NP_000320.1 | ||
| XM_017002027.1 | XP_016857516.1 | |||
| AK289925.1 | ||||
| BC075035.2 | AAH75035.1 | |||
| BC075036.2 | AAH75036.1 | |||
| U18991.1 | AAA99012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546450 | Chr.1:68405344 - 68466239 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map