Assay Details
Target Gene Details
Entrez Gene ID: | 100526835 |
Gene Name: | FPGT-TNNI3K readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:74198212-74544432 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FPGT-TNNI3K | NM_001112808.2 | NP_001106279.2 | ||
| NM_001199327.1 | NP_001186256.2 | |||
| AB208941.1 | BAD92178.1 | |||
| BC032865.1 | AAH32865.1 | |||
| BX640903.1 | CAE45949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893723 | Chr.1:73643288 - 74246497 on Build GRCh38 | Gain |
|
| nsv1002115 | Chr.1:72379872 - 74933684 on Build GRCh38 | Gain |
|
| nsv522147 | Chr.1:73872847 - 74518151 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map