Assay Details
Target Gene Details
Entrez Gene ID: | 116461 |
Gene Name: | tRNA splicing endonuclease subunit 15 |
Gene Aliases: |
C1orf19, PCH2F, sen15 |
Location: |
Chr.1:184050207-184095846 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSEN15 | XM_011509139.2 | XP_011507441.1 | ||
| XM_017000228.1 | XP_016855717.1 | |||
| XM_017000229.1 | XP_016855718.1 | |||
| XM_017000230.1 | XP_016855719.1 |
More Information
Set Membership: |
|

Genomic Map