Assay Details
Target Gene Details
Entrez Gene ID: | 10465 |
Gene Name: | peptidylprolyl isomerase H |
Gene Aliases: |
CYP-20, CYPH, SnuCyp-20, USA-CYP |
Location: |
Chr.1:42657770-42681654 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIH | NM_006347.3 | NP_006338.1 | ||
| XM_005270362.1 | XP_005270419.1 | |||
| XM_005270366.3 | XP_005270423.1 | |||
| XM_011540508.1 | XP_011538810.1 | |||
| XM_017000057.1 | XP_016855546.1 | |||
| XM_017000058.1 | XP_016855547.1 | |||
| XM_017000059.1 | XP_016855548.1 | |||
| AF016371.1 | AAC51927.1 | |||
| AF036331.1 | AAC60793.1 | |||
| AK294288.1 | ||||
| AK309438.1 | ||||
| AK311979.1 | ||||
| BC003412.1 | AAH03412.1 | |||
| BT019343.1 | AAV38150.1 | |||
| BU955702.1 | ||||
| CR541899.1 | CAG46697.1 | |||
| CR541920.1 | CAG46718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577249 | Chr.1:42576524 - 42885152 on Build GRCh38 | Gain |
|
| nsv829682 | Chr.1:42589429 - 42773568 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map