Assay Details
Target Gene Details
Entrez Gene ID: | 6045 |
Gene Name: | ring finger protein 2 |
Gene Aliases: |
BAP-1, BAP1, DING, HIPI3, RING1B, RING2 |
Location: |
Chr.1:185045419-185102608 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF2 | NM_007212.3 | NP_009143.1 | ||
| XM_005245413.3 | XP_005245470.1 | |||
| XM_011509852.2 | XP_011508154.1 | |||
| AF141327.1 | AAD29717.1 | |||
| AK091574.1 | ||||
| AK314793.1 | ||||
| BC012583.1 | AAH12583.1 | |||
| BX338357.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525731 | Chr.1:184843031 - 185068972 on Build GRCh38 | Gain |
|
| esv3588134 | Chr.1:185064271 - 185073474 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map