Assay Details
Target Gene Details
Entrez Gene ID: | 7107 |
Gene Name: | G protein-coupled receptor 137B |
Gene Aliases: |
TM7SF1 |
Location: |
Chr.1:236142502-236211059 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR137B | NM_003272.3 | NP_003263.1 | ||
| XM_017002209.1 | XP_016857698.1 | |||
| XM_017002210.1 | XP_016857699.1 | |||
| AF027826.1 | AAC39669.1 | |||
| AK223632.1 | BAD97352.1 | |||
| AK295408.1 | ||||
| AK301552.1 | ||||
| AK312908.1 | ||||
| AY242135.1 | AAO92302.1 | |||
| BC012100.1 | AAH12100.1 | |||
| CR541770.1 | CAG46569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv590n100 | Chr.1:236139422 - 236267713 on Build GRCh38 | Gain |
|
| nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain |
|
| nsv549397 | Chr.1:236121000 - 236194091 on Build GRCh38 | Loss |
|
| nsv832969 | Chr.1:236111012 - 236311443 on Build GRCh38 | Gain |
|
| nsv549394 | Chr.1:235876577 - 236629706 on Build GRCh38 | Gain |
|
| nsv1012436 | Chr.1:235860657 - 236648100 on Build GRCh38 | Loss |
|

Genomic Map