Assay Details
Target Gene Details
Entrez Gene ID: | 51107 |
Gene Name: | aph-1 homolog A, gamma-secretase subunit |
Gene Aliases: |
6530402N02Rik, APH-1, APH-1A, CGI-78 |
Location: |
Chr.1:150265399-150282291 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APH1A | XM_017001417.1 | XP_016856906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009508 | Chr.1:150238763 - 150298405 on Build GRCh38 | Loss |
|
| nsv1011223 | Chr.1:150256810 - 150306949 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map