Assay Details
Target Gene Details
Entrez Gene ID: | 255738 |
Gene Name: | proprotein convertase subtilisin/kexin type 9 |
Gene Aliases: |
FH3, HCHOLA3, LDLCQ1, NARC-1, NARC1, PC9 |
Location: |
Chr.1:55039476-55064853 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCSK9 | NM_174936.3 | NP_777596.2 | ||
| NR_110451.1 | ||||
| AK075365.1 | BAC11572.1 | |||
| AK122717.1 | ||||
| AK124635.1 | BAC85910.1 | |||
| AK297473.1 | ||||
| EF692496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517995 | Chr.1:55058311 - 55060623 on Build GRCh38 | Loss |
|
| nsv518638 | Chr.1:55044199 - 55060623 on Build GRCh38 | Loss |
|
| nsv518539 | Chr.1:55034756 - 55087881 on Build GRCh38 | Loss |
|
| nsv527707 | Chr.1:55050331 - 55060623 on Build GRCh38 | Loss |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map