Assay Details
Target Gene Details
Entrez Gene ID: | 55277 |
Gene Name: | FGGY carbohydrate kinase domain containing |
Gene Aliases: |
- |
Location: |
Chr.1:59296405-59762730 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGGY | NM_001113411.1 | NP_001106882.1 | ||
| NM_018291.3 | NP_060761.3 | |||
| XM_011541730.1 | XP_011540032.1 | |||
| XM_011541731.1 | XP_011540033.1 | |||
| XM_017001645.1 | XP_016857134.1 | |||
| XM_017001651.1 | XP_016857140.1 | |||
| XM_017001655.1 | XP_016857144.1 | |||
| XM_017001656.1 | XP_016857145.1 | |||
| XM_017001659.1 | XP_016857148.1 | |||
| XM_017001663.1 | XP_016857152.1 | |||
| XM_017001665.1 | XP_016857154.1 | |||
| XM_017001671.1 | XP_016857160.1 | |||
| XM_017001672.1 | XP_016857161.1 | |||
| XM_017001674.1 | XP_016857163.1 | |||
| XM_017001675.1 | XP_016857164.1 | |||
| DA969344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829982 | Chr.1:59247176 - 59367120 on Build GRCh38 | Loss |
|
| esv2748874 | Chr.1:59296403 - 59296956 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map