Assay Details
Target Gene Details
Entrez Gene ID: | 7175 |
Gene Name: | translocated promoter region, nuclear basket protein |
Gene Aliases: |
- |
Location: |
Chr.1:186311652-186375325 on Build GRCh38 |
Assay Gene Location: | Within Intron 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPR | NM_003292.2 | NP_003283.2 | ||
| U69668.1 | AAB48030.1 | |||
| X66397.1 | CAA47021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575361 | Chr.1:186266048 - 186604612 on Build GRCh38 | Gain |
|
| esv3890826 | Chr.1:185730915 - 186448352 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map