Assay Details
Target Gene Details
Entrez Gene ID: | 100506023 |
Gene Name: | uncharacterized LOC100506023 |
Gene Aliases: |
- |
Location: |
Chr.1:173235060-173477155 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506023 | NR_037845.1 | |||
| AK127238.1 |
Target Gene Details
Entrez Gene ID: | 7292 |
Gene Name: | tumor necrosis factor superfamily member 4 |
Gene Aliases: |
CD134L, CD252, GP34, OX-40L, OX4OL, TNLG2B, TXGP1 |
Location: |
Chr.1:173183729-173462208 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF4 | XM_017002229.1 | XP_016857718.1 | ||
| XM_017002230.1 | XP_016857719.1 |
More Information
Set Membership: |
|

Genomic Map