Assay Details
Target Gene Details
Entrez Gene ID: | 1629 |
Gene Name: | dihydrolipoamide branched chain transacylase E2 |
Gene Aliases: |
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B |
Location: |
Chr.1:100186922-100249864 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DBT | NM_001918.3 | NP_001909.3 | ||
| XM_005270545.3 | XP_005270602.1 | |||
| XM_017000468.1 | XP_016855957.1 | |||
| XM_017000469.1 | XP_016855958.1 | |||
| AK303941.1 | ||||
| AK313191.1 | ||||
| BC016675.1 | AAH16675.1 | |||
| BT007372.1 | AAP36036.1 | |||
| J03208.1 | AAA35589.1 | |||
| M27093.1 | AAA64512.1 | |||
| X66785.1 | CAA47285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map