Assay Details
Target Gene Details
Entrez Gene ID: | 5792 |
Gene Name: | protein tyrosine phosphatase, receptor type F |
Gene Aliases: |
BNAH2, LAR |
Location: |
Chr.1:43522238-43623672 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRF | XM_006710795.3 | XP_006710858.1 | ||
| XM_011541871.2 | XP_011540173.1 | |||
| XM_011541872.2 | XP_011540174.1 | |||
| XM_011541873.2 | XP_011540175.1 | |||
| XM_017001949.1 | XP_016857438.1 | |||
| DA339772.1 | ||||
| KM111191.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv157n100 | Chr.1:43525952 - 43606749 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map