Assay Details
Target Gene Details
Entrez Gene ID: | 55277 |
Gene Name: | FGGY carbohydrate kinase domain containing |
Gene Aliases: |
- |
Location: |
Chr.1:59296405-59762730 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3586180 | Chr.1:59319533 - 59367279 on Build GRCh38 | Loss |
|
| nsv829982 | Chr.1:59247176 - 59367120 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76241027] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map