Assay Details
Target Gene Details
Entrez Gene ID: | 4725 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit S5 |
Gene Aliases: |
CI-15k, CI15K |
Location: |
Chr.1:39026295-39034636 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFS5 | NM_001184979.1 | NP_001171908.1 | ||
NM_004552.2 | NP_004543.1 | |||
AF020352.1 | AAB87866.1 | |||
AF047434.1 | AAC39878.1 | |||
AF086268.1 | ||||
AK312188.1 | ||||
BC001884.1 | AAH01884.1 | |||
BC093057.1 | AAH93057.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3585778 | Chr.1:39021596 - 39033061 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
