Assay Details
Target Gene Details
Entrez Gene ID: | 5733 |
Gene Name: | prostaglandin E receptor 3 |
Gene Aliases: |
EP3, EP3-I, EP3-II, EP3-III, EP3-IV, EP3-VI, EP3e, PGE2-R |
Location: |
Chr.1:70852353-71047814 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGER3 | NM_198714.1 | NP_942007.1 | ||
| NM_198716.1 | NP_942009.1 | |||
| NM_198717.1 | NP_942010.1 | |||
| NR_028292.1 | ||||
| NR_028293.1 | ||||
| NR_028294.1 | ||||
| XM_011541810.2 | XP_011540112.1 | |||
| AK290634.1 | ||||
| AY387856.1 | AAQ90021.1 | |||
| AY429108.1 | AAR07903.1 | |||
| BC024229.1 | AAH24229.1 | |||
| D38298.1 | BAA07417.1 | |||
| D38300.1 | BAA07419.1 | |||
| D38301.1 | BAA07420.1 | |||
| D86097.1 | BAA19958.1 | |||
| D86098.1 | BAA19959.1 | |||
| L27489.1 | AAC13373.1 | |||
| S69326.1 | AAB30208.1 | |||
| U13214.1 | AAA21130.1 | |||
| U13215.1 | AAA21131.1 | |||
| X83857.1 | CAA58737.1 | |||
| X83859.1 | CAA58739.1 | |||
| X83861.1 | CAA58741.1 | |||
| X83863.1 | CAA58743.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577913 | Chr.1:70852393 - 70874208 on Build GRCh38 | Loss |
|
| nsv830148 | Chr.1:70844683 - 71018249 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map