Assay Details
Target Gene Details
Entrez Gene ID: | 84871 |
Gene Name: | ATP/GTP binding protein like 4 |
Gene Aliases: |
CCP6 |
Location: |
Chr.1:48520157-50024011 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL4 | NM_001323573.1 | NP_001310502.1 | ||
| NM_001323574.1 | NP_001310503.1 | |||
| NM_001323575.1 | NP_001310504.1 | |||
| NM_032785.3 | NP_116174.3 | |||
| NR_136623.1 | ||||
| XM_011542308.2 | XP_011540610.1 | |||
| XM_011542310.2 | XP_011540612.1 | |||
| XM_017002595.1 | XP_016858084.1 | |||
| XM_017002596.1 | XP_016858085.1 | |||
| XM_017002597.1 | XP_016858086.1 | |||
| XM_017002598.1 | XP_016858087.1 | |||
| AK094826.1 | ||||
| AK294394.1 | ||||
| AK294633.1 | ||||
| AK296221.1 |
Target Gene Details
Entrez Gene ID: | 101929721 |
Gene Name: | uncharacterized LOC101929721 |
Gene Aliases: |
- |
Location: |
Chr.1:49257411-49269294 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929721 | NR_125988.1 | |||
| AK131006.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011119 | Chr.1:49177942 - 49289300 on Build GRCh38 | Loss |
|
| nsv546194 | Chr.1:49097909 - 49583818 on Build GRCh38 | Loss |
|
| esv3577841 | Chr.1:49011673 - 49272610 on Build GRCh38 | Loss |
|
| nsv546195 | Chr.1:49238414 - 49388176 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map