Assay Details
Target Gene Details
Entrez Gene ID: | 84871 |
Gene Name: | ATP/GTP binding protein like 4 |
Gene Aliases: |
CCP6 |
Location: |
Chr.1:48520157-50024011 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL4 | NM_001323573.1 | 13 | 2390 | NP_001310502.1 |
| NM_001323574.1 | 14 | 2417 | NP_001310503.1 | |
| NM_001323575.1 | 13 | 2354 | NP_001310504.1 | |
| NM_032785.3 | 14 | 2381 | NP_116174.3 | |
| NR_136623.1 | 13 | 2353 | ||
| XM_011542308.2 | 14 | 2449 | XP_011540610.1 | |
| XM_017002595.1 | XP_016858084.1 | |||
| XM_017002597.1 | 14 | 2468 | XP_016858086.1 | |
| AK094826.1 | 14 | 2340 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585970 | Chr.1:48459872 - 48603418 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map