Assay Details
Target Gene Details
Entrez Gene ID: | 51072 |
Gene Name: | mediator of cell motility 1 |
Gene Aliases: |
C2orf4, CGI-27, MEMO, NS5ATP7 |
Location: |
Chr.2:31867188-32011052 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEMO1 | XM_011532896.2 | 7 | 1761 | XP_011531198.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv963219 | Chr.2:31865758 - 31868548 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map