Assay Details
Target Gene Details
Entrez Gene ID: | 2825 |
Gene Name: | G protein-coupled receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.2:206175316-206218047 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR1 | NM_001098199.1 | NP_001091669.1 | ||
| NM_001261452.1 | NP_001248381.1 | |||
| NM_001261453.1 | NP_001248382.1 | |||
| NM_005279.3 | NP_005270.2 | |||
| XM_017003832.1 | 1 | 68 | XP_016859321.1 | |
| AK290976.1 | ||||
| AK312917.1 | ||||
| BC067833.1 | AAH67833.1 | |||
| CB269800.1 | ||||
| DA545892.1 | ||||
| DA988428.1 |
Target Gene Details
Entrez Gene ID: | 101669764 |
Gene Name: | GPR1 antisense RNA |
Gene Aliases: |
GPR1-AS1, GPR1AS |
Location: |
Chr.2:206203376-206266243 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR1-AS | NR_104359.1 | |||
| AB774455.1 | ||||
| BE734382.1 |
More Information
Set Membership: |
|

Genomic Map