Assay Details
Target Gene Details
Entrez Gene ID: | 5610 |
Gene Name: | eukaryotic translation initiation factor 2 alpha kinase 2 |
Gene Aliases: |
EIF2AK1, PKR, PPP1R83, PRKR |
Location: |
Chr.2:37099210-37157065 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2AK2 | NM_001135651.2 | 17 | 3396 | NP_001129123.1 |
| NM_001135652.2 | 14 | 2966 | NP_001129124.1 | |
| NM_002759.3 | 17 | 3630 | NP_002750.1 | |
| XM_011532987.2 | 17 | 3330 | XP_011531289.1 | |
| XM_017004503.1 | 16 | 3197 | XP_016859992.1 | |
| BU687340.1 |
More Information
Set Membership: |
|

Genomic Map