Assay Details
Target Gene Details
Entrez Gene ID: | 103752554 |
Gene Name: | MYCN upstream transcript (non-protein coding) |
Gene Aliases: |
MYCNUN, lncUSMycN |
Location: |
Chr.2:15920399-15936018 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCNUT | NR_125783.1 | |||
| W87755.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73915542] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map