Assay Details
Target Gene Details
Entrez Gene ID: | 104326052 |
Gene Name: | negative regulator of interferon response (non-protein coding) |
Gene Aliases: |
lncRNA-CMPK2 |
Location: |
Chr.2:6828554-6840464 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRIR | NR_126359.1 | 3 | 372 | |
| BP421873.1 | 3 | 372 | ||
| BQ437827.1 | 2 | 299 |

Genomic Map