Assay Details
Target Gene Details
Entrez Gene ID: | 55233 |
Gene Name: | MOB kinase activator 1A |
Gene Aliases: |
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B |
Location: |
Chr.2:74152598-74178967 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB1A | NM_001317110.1 | NP_001304039.1 | ||
| NM_001317111.1 | NP_001304040.1 | |||
| NM_001317112.1 | NP_001304041.1 | |||
| NM_018221.4 | NP_060691.2 | |||
| XM_011532948.2 | XP_011531250.1 | |||
| AB016839.1 | BAB19058.1 | |||
| AJ577474.1 | CAE12093.1 | |||
| AK001650.1 | BAA91810.1 | |||
| AK023321.1 | BAB14525.1 | |||
| BC003398.1 | AAH03398.1 | |||
| BI825399.1 | ||||
| BU172616.1 | ||||
| CN426611.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs150104788] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map