Assay Details
Target Gene Details
Entrez Gene ID: | 2863 |
Gene Name: | G protein-coupled receptor 39 |
Gene Aliases: |
- |
Location: |
Chr.2:132415972-132646596 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR39 | NM_001508.2 | NP_001499.1 | ||
| AF034633.1 | AAC26082.1 | |||
| AK122643.1 | ||||
| AK314895.1 | ||||
| BC125046.1 | ||||
| BC125047.1 | ||||
| FJ348258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834378 | Chr.2:132412223 - 132591833 on Build GRCh38 | Loss |
|
| dgv4081n100 | Chr.2:131956759 - 132601004 on Build GRCh38 | Gain |
|
| nsv583153 | Chr.2:132409690 - 132603709 on Build GRCh38 | Gain |
|
| nsv526029 | Chr.2:131975708 - 132593842 on Build GRCh38 | Gain |
|
| dgv7085n54 | Chr.2:132276581 - 132594130 on Build GRCh38 | Gain |
|
| nsv583152 | Chr.2:132369502 - 133631550 on Build GRCh38 | Gain |
|
| nsv1011421 | Chr.2:132137668 - 132924612 on Build GRCh38 | Gain |
|
| esv2763165 | Chr.2:131961977 - 132601016 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72999271,rs73955697] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map