Assay Details
Target Gene Details
Entrez Gene ID: | 10018 |
Gene Name: | BCL2 like 11 |
Gene Aliases: |
BAM, BIM, BOD |
Location: |
Chr.2:111120914-111168445 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829618 | Chr.2:111118050 - 111123970 on Build GRCh38 | Gain |
|
| nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss |
|
| nsv582699 | Chr.2:110763166 - 112172487 on Build GRCh38 | Loss |
|
| nsv834331 | Chr.2:111054562 - 111247101 on Build GRCh38 | Loss |
|
| dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map